Imprimer
EasySep™ Mouse Naïve CD8+ T Cell Isolation Kits
Fournisseur: STEMCELL Technologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
19858 19858RF
STMC19858EA 1580 EUR
STMC19858 STMC19858RF
EasySep™ Mouse Naïve CD8+ T Cell Isolation Kits
Milieux de séparation cellulaire
Immunomagnetic isolation of untouched mouse naïve CD8+ T cells
  • Fast and easy-to-use
  • Up to 98% purity
  • No columns required
  • Untouched, viable cells
The EasySep™ Mouse Naïve CD8+ T Cell Isolation Kit is designed to isolate naïve CD62L+CD44-CD8+ T cells from single-cell suspensions of splenocytes or lymph nodes by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies that are directed against non-naïve CD8+ T cells (CD4, CD11b, CD11c, CD19, CD44, CD45R/B220, CD49b, TCRγ/δ, TER119) and streptavidin-coated magnetic particles. labelled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.
Attention: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').