Imprimer
EasySep™ Mouse Pan-DC Enrichment Kit II
Fournisseur: STEMCELL Technologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
19863 19863RF
STMC19863EA 1610 EUR
STMC19863 STMC19863RF
EasySep™ Mouse Pan-DC Enrichment Kit II
Milieux de séparation cellulaire
Immunomagnetic negative selection kit
  • Fast, easy-to-use, and column-free
  • Up to 65% purity
  • Isolated cells are untouched
  • Optimized for high yield
  • Facilitates rapid flow sorting of DC subsets
The EasySep™ Mouse Pan-DC Enrichment Kit II is designed to isolate dendritic cells (DCs) from mouse splenocytes by immunomagnetic negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labelled cells are separated from the untouched desired cells using an EasySep™ magnet and simplyEn savoir plus