Imprimer
EasySep™ HLA B Cell Enrichment: Complete Processing Kits for Whole Blood
Fournisseur: STEMCELL Technologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
19954HLA 19954HLARF
STMC19954HLAEA 1100 EUR
STMC19954HLA STMC19954HLARF
EasySep™ HLA B Cell Enrichment: Complete Processing Kits for Whole Blood
Milieux de séparation cellulaire
Immunomagnetic isolation of untouched human B cells
  • Fast, easy-to-use and column-free
  • Up to 99% purity
  • Untouched, viable cells
  • Compatible with downstream HLA assays
The EasySep™ HLA B Cell Enrichment Kit: Complete Processing Kit for Whole Blood is designed to isolate human B cells from HetaSep™-treated whole blood by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognising CD2, CD3, CD14, CD16, CD33, CD36, CD41, CD43, CD56, CD66b, glycophorin A and dextran-coated magnetic particles. The labelled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.
Attention: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').