Imprimer
RosetteSep™ CTC Enrichment Cocktail Containing Anti-CD56s
Fournisseur: STEMCELL Technologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
15137 15177
STMC15137EA 910 EUR
STMC15137 STMC15177
RosetteSep™ CTC Enrichment Cocktail Containing Anti-CD56s
Milieux de séparation cellulaire
Immunodensity isolation of untouched circulating tumor cells.
  • Fast and easy-to-use
  • Requires no special equipment or training
  • Untouched, viable cells
  • Can be combined with SepMate™ for consistent, high-throughput sample processing
The RosetteSep™ CTC Enrichment Cocktail Containing Anti-CD56 is designed to enrich circulating epithelial tumor cells from fresh whole blood by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognising CD3, CD14, CD16, CD19, CD38, CD45, CD56, CD61, CD66b and glycophorin A on red blood cells (RBCs). When centrifuged over a density gradient medium such as Lymphoprep™ (STMC07801), the unwanted cells pellet along with the RBCs. The purified tumor cells are present as a highly enriched population at the interface between the plasma and the density gradient medium.
This cocktail is compatible with breast cancer samples. If you are working with small-cell carcinoma samples, including lung cancer samples, use the RosetteSep™ CTC Enrichment Cocktail Containing Anti-CD36 (STMC15127).
Attention: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).