Imprimer
RosetteSep™ Human CD8 Depletion Cocktails
Fournisseur: STEMCELL Technologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
15623 15663
STMC15623EA 226 EUR
STMC15623 STMC15663
RosetteSep™ Human CD8 Depletion Cocktails
Milieux de séparation cellulaire
Immunodensity depletion of CD8+ T cells.
  • Fast and easy-to-use
  • Requires no special equipment or training
  • Untouched, viable cells
  • Can be combined with SepMate™ for consistent, high-throughput sample processing
The RosetteSep™ Human CD8 Depletion Cocktail is designed to deplete CD8+ cells from whole blood. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognising CD8 and glycophorin A on red blood cells (RBCs). When centrifuged over a buoyant density medium such as Lymphoprep™ (STMC07801), the unwanted cells pellet along with the RBCs. The CD8+ depleted fraction is present as a highly enriched population at the interface between the plasma and the buoyant density medium.
Attention: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).