Imprimer
RosetteSep™ HLA Granulocyte Depletion Cocktails
Fournisseur: STEMCELL Technologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
15684HLA 15664HLA
STMC15684HLAEA 4730 EUR
STMC15684HLA STMC15664HLA
RosetteSep™ HLA Granulocyte Depletion Cocktails
Milieux de séparation cellulaire
Immunodensity depletion of granulocytes for HLA analysis.
  • Fast and easy-to-use
  • Requires no special equipment or training
  • Isolated cells are untouched
The RosetteSep™ HLA Granulocyte Depletion Cocktail is designed to deplete granulocytes from whole blood. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognising CD66b and glycophorin A on red blood cells (RBCs). When centrifuged over a buoyant density medium such as Lymphoprep™ (STMC07801), the unwanted cells pellet along with the RBCs. The desired cells are present as a highly enriched population depleted of granulocytes at the interface between the plasma and the buoyant density medium.
Please Note: In light of the new EU Regulation 2017/746 (IVDR) of the European Parliament and of the Council on in vitro diagnostic medical devices, which is expected to become effective in 2022, STEMCELL recently deregistered and removed the CE IVD claims associated with these products, and they are now labeled 'For Research Use Only'. Please note there have been no changes to the form, function and quality assurances associated with these products.
Attention: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).