Imprimer
EasySep™ Mouse Biotin Positive Selection Kit II
Fournisseur: STEMCELL Technologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
17665 17665RF
STMC17665EA 978 EUR
STMC17665 STMC17665RF
EasySep™ Mouse Biotin Positive Selection Kit II
Cell Separation Media
Immunomagnetic positive selection of mouse cells labeled with biotinylated antibodies.
  • Fast and easy-to-use
  • No columns required
The EasySep™ Mouse Biotin Positive Selection Kit II is designed to isolate cells that are labeled with biotinylated antibodies. Desired cells are targeted with antibody complexes recognising biotin and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.
This product replaces the EasySep™ Mouse Biotin Positive Selection Kit.
Attention: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).