Imprimer
EasySep™ Human CD4 Positive Selection Kit II
Fournisseur: STEMCELL Technologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
17852RF 17852 100-0693
STMC17852RFEA 1390 EUR
STMC17852RF STMC17852 STMC100-0693
EasySep™ Human CD4 Positive Selection Kit II
Milieux de séparation cellulaire
Immunomagnetic positive selection of human CD4+ cells (e.g. CD4+ T cells).
  • Fast and easy-to-use
  • Up to 96% purity
  • No columns required
The EasySep™ Human CD4 Positive Selection Kit II is designed to isolate CD4+ cells from fresh or previously frozen peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic positive selection. Desired cells are targeted with antibody complexes recognising CD4 and magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.
This product replaces the EasySep™ Human CD4 Positive Selection Kit (STMC18052) for even faster cell isolations.
Attention: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).