Imprimer
EasySep™ Human CD19 Positive Selection Kit II
Fournisseur: STEMCELL Technologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
17854 17854RF
STMC17854EA 1300 EUR
STMC17854 STMC17854RF
EasySep™ Human CD19 Positive Selection Kit II
Milieux de séparation cellulaire
Immunomagnetic positive selection of human CD19+ cells (e.g. B cells).
  • Fast and easy-to-use
  • Up to 99% purity
  • No columns required
The EasySep™ Human CD19 Positive Selection Kit II is designed to isolate CD19+ cells from fresh or previously frozen peripheral blood mononuclear cells by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognising CD19 and dextran-coated magnetic particles. The cocktail also contains an antibody to human Fc receptor to minimise nonspecific binding. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD19 antigen is expressed on mature B cells and B cell progenitors but is lost upon maturation to plasma cells. It is also expressed on follicular dendritic cells.
This product replaces the EasySep™ Human CD19 Positive Selection Kit (STMC18054) for even faster cell isolations.
Attention: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).