Imprimer
EasySep™ HLA Chimerism Whole Blood Lymphoid Positive Selection Kits
Fournisseur: STEMCELL Technologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
17873 17873RF
STMC17873EA 1350 EUR
STMC17873 STMC17873RF
EasySep™ HLA Chimerism Whole Blood Lymphoid Positive Selection Kits
Milieux de séparation cellulaire
Column-free immunomagnetic positive selection of human CD3+ and CD19+ cells (e.g. lymphocytes) cells for HLA analysis.
  • Fast and easy-to-use
  • Up to 99.9% purity
  • No columns required
The EasySep™ Human Whole Blood Lymphoid Positive Selection Kit is designed to isolate CD3+ and CD19+ cells from whole blood or buffy coat by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognising CD3, CD19 and dextran-coated magnetic particles. The cocktail also contains an antibody to human Fc receptor to minimise nonspecific binding.En savoir plus