Imprimer
Full Facepiece Reusable Respirators, FF-600
Fournisseur: 3M
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
FM4-M FM4-S
MMMAFM4-MEA 472 EUR
MMMAFM4-M MMMAFM4-S
Full Facepiece Reusable Respirators, FF-600
Protection respiratoire Masques réutilisables Masques complets
The 3M™ full facepiece reusable respirator FF-600 series can be used with approved 3M™ DT-Series filters with a 40 mm (EN 148-1) thread, or they can be used as part of an approved powered air respirator system using the appropriate breathing tube. Both combinations protect against gases, vapours and/or particulates depending on your individual needs.
  • Speech diaphragm
  • Side port filter or breathing tube attachment
  • Three sizes FF-601 (small) FF-602 (medium) FF-603 (medium/large)
  • Spectacle kit available
  • Facepiece weight: approx. 550 g
Please note, the 3M™ reusable full face mask, FF-600, was previously named the Scott™ vision two reusable full face mask.
Certifications: The 3M™ full facepiece reusable respirator FF-600 series have been tested to EN 136:1998 Class 2 and EN 12942:1998 + A2:2008 TM3.