Imprimer
Multi-Cassette (Urine)
Fournisseur: SURESCREEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
S12MC G10MC2300
SURES12MCEA 288 EUR
SURES12MC SUREG10MC2300
Multi-Cassette (Urine)
Drug Tests
The Multi-Cassette only requires a small amount of urine to perform the test, with test results available at 5 minutes. This multiline format test comes with a pipette for an easy-to-use process.
  • Multi-drug test incorporated
  • Suitable for low-volume sample
  • Test up to 12 parameters simultaneously