Imprimer
Intelsius® BioTherm™ Dry Ice Shippers
Fournisseur: DGP INTELSIUS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BT074 BT041 BT049 BT020 BT050
DGINBT074EA 18.5 EUR
DGINBT074 DGINBT041 DGINBT049 DGINBT020 DGINBT050
Intelsius® BioTherm™ Dry Ice Shippers
Boîtes d'expédition et de transport
Intelsius® Biotherm packaging systems are regulatory compliant to IATA, ADR, and 49 CFR (DOT) transport regulations. These shippers utilise dry ice to maintain sample integrity during shipments from 24 to 240 hours.
  • Regulatory compliant
  • Dry ice shippers
  • Compliant markings on boxes
  • Variety of internal volumes
Intelsius® Biotherm shipping systems are pre-qualified to maintain sample integrity during shipments from 24 to 240 hours against demanding high-performance dry ice temperature (2 to 8 °C profiles and 15 to 25 °C profiles). The range is tested and qualified in conjunction with Intelsius® Specimen transport packaging.
The range is designed to optimise shipping volume and minimize freight costs. These insulated shipping systems have unique, sturdy carry handles to enable careful handling throughout the cold chain and a tongue and groove lid and base to enable quick and simple assembly. Pack-out instruction sheets are supplied to ensure correct assembly.
Certifications: Class 6.2, Category A and B.