Imprimer
, en poudre
Marques : MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s): Phosphate de disodium et de 4-nitrophényle hexahydraté, pNPP
0210087825 0210087850 02100878.5 0210087801 0210087805 0210087810
ICNA0210087825EA 249 EUR
ICNA0210087825 ICNA0210087850 ICNA02100878.5 ICNA0210087801 ICNA0210087805 ICNA0210087810
, en poudre
Nitro-4 phényle phosphate sel de sodium hexahydraté (pNPP)
p nitrophenol is the substrate of choice for use with alkaline phosphatase in ELISA procedures. This substrate produces a soluble end product that is yellow in color and can be read spectrophotometrically at 405 nm. The pNPP reaction may be stopped with 3 M NaOH and read at 405 nm.
p-nitrophenol is suitable for use as a substrate for alkaline and acid phosphatase.
Typically a pNPP concentration of 1 mg/ml is used
Soluble in water.
Formule: C₆H₄NNa₂O₆P·6H₂O
Poids moléculaire: 371.144 g/mol
Point de fusion: >300 °C (lit.)                      
Température de stockage: Congélateur
Numéro MDL: MFCD00066288
Numéro CAS: 333338-18-4
EINECS: 224-246-5
Commander maintenant
Résultats des tests de spécification
Identity Test Passes
Water Content 27 - 31%
Free p-Nitrophenol ≤0,005%