Imprimer
Volumetric Flasks, Class A/B, PMP, NS Plastic Stopper, One Batch
Fournisseur: Brand
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2.36174 2.36176
BRND2.36174EA 0 EUR
BRND2.36174 BRND2.36176
Volumetric Flasks, Class A/B, PMP, NS Plastic Stopper, One Batch
Ballons Fioles jaugées
PMP, transparent, with PP stopper.
  • Calibrated to contain (TC, In)
  • Ring mark and inscriptions in blue
Error limits according to class A respectively B, DIN EN ISO 1042.
Exposure to temperature up to 121 °C (autoclaving) will not cause permanently exceeded error limits! Cleaning up to 60 °C is recommended to preserve marks and inscriptions.