Imprimer
AlphaTec® 4000, 132 Chemical Resistant Overalls
Fournisseur: Ansell
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
GR40T-00132-01 GR40T-00132-02 GR40T-00132-03 GR40T-00132-04 GR40T-00132-05 GR40T-00132-06 GR40T-00132-07 GR40T-00132-08 GR40T-00132-09
ANSEGR40T-00132-01EA 609 EUR
ANSEGR40T-00132-01 ANSEGR40T-00132-02 ANSEGR40T-00132-03 ANSEGR40T-00132-04 ANSEGR40T-00132-05 ANSEGR40T-00132-06 ANSEGR40T-00132-07 ANSEGR40T-00132-08 ANSEGR40T-00132-09
AlphaTec® 4000, 132 Chemical Resistant Overalls
Vêtements à usage unique
Innovative multi-layer chemical barrier technology, type 3/4/5 protection.
  • Over 200 chemicals permeation tested: Superb chemical protection
  • Welded and taped seams: Ensures barrier from liquids and particulates
  • Knitted inner cuffs: Offers wearers high levels of comfort
  • Coverall, 2-piece hood, single zip flap
  • Elasticated hood, waist, wrists and ankles and finger loops
Specialised protection: The AlphaTec® 4000 Ultrasonically Welded and Taped - Model 132 chemical protective suit safeguards wearers from over 200 chemicals permeation, such as chemical warfare agents.
Advanced defenses: This protective suit features welded and taped seams that act as barrier against liquids and particulates to increase personal protection.
Elevated comfort: Delivering greater comfort, this protective body suit features knitted inner cuffs and inner textile type fabric.
Certified anti-static: Tested and approved in accordance with EN 1149-5, this full chemical protection suit meets anti-static standards.
Recommended For:
Contaminated environments
Chemical handling
Filling, blending and charging of raw materials
Transferring liquids and solids
Opening and draining pumps valves or lines
Loading and unloading of proces equipment
Raw material sample collection
Concentration mixing and palletisation
Hazmat response
Assembly and disassembly of equipment
Primary industries: Chemical, life sciences, mining, fire and rescue, oil and gas, automotive aftermarket, food processing.
Material: Multi-layer non-woven barrier laminate fabric
Shelf life: Five years