Imprimer
BD Emerald™ Syringes
Fournisseur: BD Medical
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
307727 307736 307731 303126 303127 303128 307742
613-2943EA 27.6 EUR
613-2943 BDAM307736 BDAM307731 BDAM303126 BDAM303127 BDAM303128 BDAM307742
BD Emerald™ Syringes
Seringues Seringues médicales
Sterile, disposable, three-piece syringes, PP barrel and piston, latex-free stopper. Luer slip syringes available with or without needle. Combining high-quality performance and reduced environmental impact.
  • For general purpose use. Specialised design features benefit clinicians, patients and the environment
  • Green stopper and bold scale markings to aid in setting an accurate dose
  • Clear barrel allows a clear view of fluid
  • Retaining ring to help prevent accidental plunger rod pullout
  • Flange size and shape to provide stability and comfort during use
  • Textured thumb press to reduce slippage during administration
  • Minimises waste disposal - has up to 30% less material than other syringes
  • Sterile, single use
  • Natural rubber latex is not part of the material formulation
Attention: BD Emerald™ 3-piece syringes are not designed for use on syringe pumps.