Imprimer
EasySep™ EasyStand™ Linkable Magnet Stands
Fournisseur: STEMCELL Technologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
18134 18136 18130
STMC18134EA 0 EUR
STMC18134 STMC18136 STMC18130
EasySep™ EasyStand™ Linkable Magnet Stands
Aimants
Linkable stands for EasySep™ magnets available in one, four, or six piece configurations.
  • Simplify the separation of one or more samples at one time
  • Magnets not included
Each EasySep™ EasyStand™ can hold a single EasySep™ magnet, and a simple system links up to 6 individual EasySep™ EasyStands™ together. After each round of magnetic separation, the supernatants from all samples are poured off simultaneously into the receiving tubes.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').