Imprimer
Brady® PermaShield™ Matte Polyester Labels with Overlaminate for BradyJet™ J7300 Colour Label Printer
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
176650 176653 176652 176649 176651
BRDY176650EA 686 EUR
BRDY176650 BRDY176653 BRDY176652 BRDY176649 BRDY176651
Brady® PermaShield™ Matte Polyester Labels with Overlaminate for BradyJet™ J7300 Colour Label Printer
Étiquettes
Upgrade the durability and longevity of identification projects, even in the toughest environments, with PermaShield™ Polyester Labels designed for BradyJet™ printers.
  • Protects against numerous harsh solvents and provides a more glossy appearance
  • Works best on flat, smooth surfaces
  • Inkjet receptive polyester material
  • Designed for indoor use
PermaShield™ by Brady is a 2-in-1 package that includes a printable label plus matching clear polyester overlaminate. Simply print label, adhere to surface, and apply the matching overlaminate. Commonly used to print product labelling, logos, 'Right to Know' labels, and barcode applications. Built-in LabelSense™ technology enables cartridge and printer communication for fast changeovers, automatic label setup, and seamless software interaction.
For use with the BradyJet™ J7300 Colour Label Printer. Ink cartridges are required to print on this material; use part numbers P-CMY-1 and P-K-1.