Imprimer
Brady® PermaShield™ Vinyl Labels with Overlaminate for BradyJet™ J7300 Colour Label Printer
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
176648 176655 176657
BRDY176648EA 318 EUR
BRDY176648 BRDY176655 BRDY176657
Brady® PermaShield™ Vinyl Labels with Overlaminate for BradyJet™ J7300 Colour Label Printer
Étiquettes
Upgrade the durability and longevity of identification projects, even in the toughest environments, with PermaShield™ Vinyl Labels for the BradyJet J7300 Printer.
  • Protects against harsh solvents, outdoor elements, and food and beverage washdowns
  • Label topcoat is specially formulated to invigorate colours and keep printed content clear and visible
  • Commonly used to print and place photo labels on lockout padlocks to keep workers better identified and protected during maintenance
  • Inkjet receptive vinyl material
  • Designed for indoor/outdoor use
PermaShield™ by Brady is a 2-in-1 package that includes a printable vinyl label plus matching clear polyester overlaminate. Simply print label, adhere to surface, and apply the matching overlaminate. Built-in LabelSense™ technology enables cartridge and printer communication for fast changeovers, automatic label setup and seamless software interaction.
For use with the BradyJet™ J7300 Colour Label Printer. Ink cartridges are required to print on this material; use part numbers P-CMY-1 and P-K-1.