Imprimer
Brady® Vinyl Labels for BradyJet™ J7300 Colour Label Printer
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
176603 176630 170459 176629 176601 176602 176627 176628 176626 176604
BRDY176603EA 371 EUR
BRDY176603 BRDY176630 BRDY170459 BRDY176629 BRDY176601 BRDY176602 BRDY176627 BRDY176628 BRDY176626 BRDY176604
Brady® Vinyl Labels for BradyJet™ J7300 Colour Label Printer
Étiquettes
General-purpose vinyl labels feature a specially formulated topcoat for improved durability and colour.
  • Low-shrink vinyl with ultra-aggressive adhesive is versatile for a wide array of applications
  • Tested to withstand exposure to liquids, alcohol and chemicals, as well as indoor fluorescent and UV lighting
  • Works well in low temperatures with a service temperature of −40 to +60° C
  • Inkjet receptive vinyl material
  • Designed for indoor/outdoor use
General-purpose vinyl labels are durable up to two years outdoors, even longer indoors. Ideal for Lean, general facility, pipe marking, OSHA/ANSI, PPE, machine labels and tags, quality and control, lockout, GHS and chemical, warehousing, general laboratory and more. Enhanced topcoat works with durable J7300 inks to protect printed text and graphics from smudging and harsh conditions. Built-in LabelSense™ technology enables cartridge and printer communication for fast changeovers, automatic label setup and seamless software interaction.
For use with the BradyJet™ J7300 Colour Label Printer. Ink cartridges are required to print on this material; use part numbers P-CMY-1 and P-K-1.