Imprimer
Paper Labels B-2550 for BradyJet™ J7300 Inkjet Printer
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
176621 176623 170472 176622
BRDY176621EA 198 EUR
BRDY176621 BRDY176623 BRDY170472 BRDY176622
Paper Labels B-2550 for BradyJet™ J7300 Inkjet Printer
Étiquettes
Low cost and high contrast, these inkjet receptive paper labels offer excellent smear resistance for economical light-duty identification. For use on smooth surfaces.
  • Permanent acrylic adhesive ensures labels adhere strongly to a variety of flat and curved surfaces
  • Inkjet receptive paper material
  • Designed for indoor use
  • Matt finish
Ideal for barcoding, quality and control (testing/production, circle labels, date/time), and warehousing racks, bins, and storage. Permanent acrylic adhesive ensures labels adhere strongly to a variety of flat and curved surfaces, including cardboard, plastic and metal. Built-in LabelSense™ technology enables cartridge and printer communication for fast changeovers, automatic label setup, and seamless software interaction.
For use with the BradyJet™ J7300 Colour Label Printer. Ink cartridges are required to print on this material; use part numbers P-CMY-1 and P-K-1.
Certifications: 2011/65/EU and 2015/863