Imprimer
Ubbelohde Viscometers (DIN), with TC Sensors
Fournisseur: SI Analytics
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
285423505 285423485
SCOI285423505EA 1910 EUR
SCOI285423505 SCOI285423485
Ubbelohde Viscometers (DIN), with TC Sensors
Viscometers Capillary Viscometers
Ubbelohde glass capillary viscometer with TC sensors.
  • For use with AVS® devices, equipped for thermoelectric detection, e.g. ViscoPump III (TC), VZ 8562 for thermo-electric detection
  • No measuring stand of AVS®/S series required
Certifications: According to DIN 53000-1, characteristics according to DIN 51562-1 and ISO 3105.