Imprimer
Alaris™ GW Series Oncology Primary Set
Fournisseur: BD Medical
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Alaris™
BDAM273-954EBEA 767 EUR
BDAM273-954EB
Alaris™ GW Series Oncology Primary Set
Perfusions
Oncology primary set with 15 µm drip chamber filter, needle-free connectors.
  • Light resistant
  • Bore 3 mm/1 spike
  • In-line drip chamber
  • Drops per ml 20
  • Hydrophobic air inlet filter (with cap)
  • Low sorbing/polyethylene/polyurethane
Set with 2 pinch clamp/1 roller clamp, silicone pump segment, 1 back-check valve, 1 male Luer, rotating male Luer, hydrophobic priming cap.