Imprimer
ReadiPrep™ Exosome Isolation Kit
Marques : AAT BIOQUEST
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
60204 60205
AATB60204EA 266 EUR
AATB60204 AATB60205
ReadiPrep™ Exosome Isolation Kit
Tests Tests cellulaires
Designed for consistent and dependable results. The ReadiPrep™ exosome isolation kit offers a streamlined and effective approach to isolating intact exosomes from diverse biological samples, including cell culture media and serum.
  • Robust protocol
  • Produced exosomes are suitable for further downstream analyses or purification via affinity-based methods
  • Six-month shelf life
The principle behind this kit is based on the selective precipitation of exosomes by inducing the removal of water molecules from the solution.
Informations de livraison: Shipped at RT.
Attention: Research use only.