Imprimer
Buccutite™ Rapid PE-Cy5 Tandem Antibody Labelling Kit
Fournisseur: AAT Bioquest
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Buccutite™
AATB5416EA 2290 EUR
AATB5416
Buccutite™ Rapid PE-Cy5 Tandem Antibody Labelling Kit
Systèmes de liaison et de marquage des protéines
Production scale optimised for labelling 1 mg antibody per reaction. Buccutite™ Rapid PE-Cy5 tandem antibody labelling kits, designed for large-scale production, provide a streamlined approach for labelling antibodies with PE, APC, PerCP and iFluor® tandem dyes.
  • Robust protocol
  • Consistent results
  • Six-month shelf life
Designed to provide high quality material for downstream fluorescent applications, including flow cytometry.
Informations de livraison: Shipped with blue ice.
Attention: Research use only.