Imprimer
Cell Counting Kit 8 (CCK-8)
Fournisseur: AAT Bioquest
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AATB22772EA 297 EUR
AATB22772 AATB22773
Cell Counting Kit 8 (CCK-8)
Tests Tests cellulaires
Designed for consistent and dependable results. Cell Counting Kit 8 (CCK-8) is a colorimetric assay used to quantitate cell proliferation, cell viability and cytotoxicity
  • Improved water solubility compared to MTT assays
  • Excellent stability allowing for long assays (24 to 48 hours)
  • Single-component, mix-and-read format
CCK-8 is a colorimetric assay that relies on production of formazan from WST-8, catalysed by endogenous cellular dehydrogenase in the presence of NADPH from metabolically active cells.
Informations de livraison: Shipped with blue ice.
Attention: Research use only.