Imprimer
Cell Meter™ Colorimetric MTS Cell Proliferation Kit
Marques : AAT BIOQUEST
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
22766 22767
AATB22766EA 433 EUR
AATB22766 AATB22767
Cell Meter™ Colorimetric MTS Cell Proliferation Kit
Tests Tests cellulaires
Designed for consistent and dependable results. The Cell Meter™ Colorimetric MTS Cell Proliferation Kit is a quick and reliable method for quantifying viable cells in proliferation and cytotoxicity assays.
  • Can be applied directly to cell culture media, bypassing the intermediate steps required by traditional MTT assays
  • Absorbance measured at 490 to 500 nm
  • Eliminates the need for washing or solubilisation steps
The Cell Meter™ Colorimetric MTS Cell Proliferation Kit is useful for evaluating cell proliferation in response to various stimuli such as growth factors, cytokines, mitogens and nutrients
Informations de livraison: Shipped with dry ice
Attention: Research use only.