Imprimer
TAQuest™ qPCR Master Mix for TaqMan Probes *High ROX*
Marques : AAT BIOQUEST
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
17286 17287
AATB17286EA 138 EUR
AATB17286 AATB17287
TAQuest™ qPCR Master Mix for TaqMan Probes *High ROX*
Réactifs pour acides nucléiques qPCR/RT-qPCR Enzymes and Kits
TAQuest™ qPCR Master mix for TaqMan probes is a ready-to-use 2X solution optimised for qPCR and two-step RT-qPCR compatible with TaqMan gene expression assays.
  • Simple and robust protocol
  • The optimised composition ensures PCR specificity and sensitivity with all sample types such as genomic, plasmid, viral and cDNA templates
TAQuest™ qPCR Master mix for TaqMan probes has been designed to be used for duplex reactions using internal positive controls.
Attention: Master mix is stable for 12 months when stored frozen (<−15 °C) with minimised light exposure.