Imprimer

Anti-IgG Goat Polyclonal Antibody (iFluor® 860)

Fournisseur: AAT BIOQUEST
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

48066 48067
AATB48066EA 138 EUR
AATB48066 AATB48067
Anti-IgG Goat Polyclonal Antibody (iFluor® 860)
Anticorps
AAT Bioquest's iFluor® dyes are optimised for labeling proteins, in particular, antibodies.

  • Purified and quality-tested for consistent results
  • Suitable for multiple applications
  • Purified through affinity chromatography

iFluor® 860 goat anti-rabbit IgG (H+L) conjugates are ideal for various NIR imaging applications, including Western blotting, ELISA, protein arrays, tissue section imaging, and in vivo imaging.

Attention: Research use only.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: iFluor® 860
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Rabbit
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR