Imprimer

iFluor® Ultra 750 succinimidyl ester

Marques : AAT BIOQUEST
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

71680 71681 71682
AATB71680EA 429 EUR
AATB71680 AATB71681 AATB71682
iFluor® Ultra 750 succinimidyl ester
iFluor® Ultra 750 succinimidyl ester

iFluor® Ultra 750 is a bright NIR fluorescent dye excitable by 633 nm to 685 nm laser lines, with a peak emission at 773 nm.


  • Stable for 12 months


Fluorescent dye-conjugated antibodies provide a tool for identifying proteins in many applications including fluorescent cell imaging, flow cytometry, western blotting, immunohistochemistry and more.


Attention: For Research Use Only

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR