Imprimer
iFluor® Ultra 647 succinimidyl ester
Marques : AAT BIOQUEST
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
71670 71671 71672
AATB71670EA 429 EUR
AATB71670 AATB71671 AATB71672
iFluor® Ultra 647 succinimidyl ester
iFluor® Ultra 647 succinimidyl ester
iFluor® Ultra 647 is an exceptionally bright far-red fluorescent dye excitable by 594 nm or 633 nm laser lines, with a peak emission at 670 nm.
  • Stable for 12 months
Fluorescent dye-conjugated antibodies provide a tool for identifying proteins in many applications including fluorescent cell imaging, flow cytometry, western blotting, immunohistochemistry and more.
Attention: For Research Use Only