iFluor® 830 succinimidyl ester
Marques : AAT BIOQUESTTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
iFluor® 830 dye emission is a unique color for spectrum flow cytometry as it is well separated from commonly used far-red fluorophores such as Cy5, Cy7, or allophycocyanin (APC), facilitating multicolor analysis.
- Stable for 12 months
In vivo fluorescence imaging uses a sensitive camera to detect the fluorescence emission from fluorophores in whole-body living small animals.
Attention: For Research Use Only