Imprimer
DBCO Sulfo-NHS ester
Marques : AAT BIOQUEST
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
4527 4528
AATB4527EA 130 EUR
AATB4527 AATB4528
DBCO Sulfo-NHS ester
DBCO Sulfo-NHS ester
DBCO-Sulfo-NHS Ester is a water-soluble version of the popular DBCO NHS Ester (#4529).
  • Stable for 12 months
Quality tested in-house
Attention: For Research Use Only