Imprimer

Anti-TIM 3 Rabbit Recombinant Antibody [clone: DM83]

Fournisseur: ANTIBODIES.COM
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

A318624-100 A318624-500
ANTIA318624-100EA 640 EUR
ANTIA318624-100 ANTIA318624-500
Anti-TIM 3 Rabbit Recombinant Antibody [clone: DM83]
Anticorps
Recombinant rabbit monoclonal [DM83] antibody to TIM 3 for ELISA and flow cytometry with samples derived from human.

  • Validated applications: ELISA, flow cytometry
  • Recommended dilutions: ELISA: 1:5000 to 10000, flow cytometry: 1:100

Type: Primary
Antigen: TIM 3
Clonality: Recombinant
Clone: DM83
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR