Imprimer

Anti-G-CSF Mouse Monoclonal Antibody [clone: KT26]

Fournisseur: ANTIBODIES.COM
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

A78-100 A78-200
ANTIA78-100EA 445 EUR
ANTIA78-100 ANTIA78-200
Anti-G-CSF Mouse Monoclonal Antibody [clone: KT26]
Anticorps
Mouse monoclonal (KT26) antibody to G-CSF for ELISA with samples derived from human.

  • Validated applications: ELISA
  • Recommended dilutions: ELISA: 1 µg/ml

Type: Primary
Antigen: G-CSF
Clonality: Monoclonal
Clone: KT26
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR