Imprimer
PlateLoc Microplate Heat Seals
Fournisseur: AGILENT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
24210-001 24214-001 06644-001 16985-001 17318-001 24212-001
AGLS24210-001EA 2149 EUR
AGLS24210-001 AGLS24214-001 AGLS06644-001 AGLS16985-001 AGLS17318-001 AGLS24212-001
PlateLoc Microplate Heat Seals
Scellés Microplaque Sceaux
Certified for use with the PlateLoc Thermal Microplate Sealer.
  • Choice of aluminum-based or optically clear materials with peelable, pierceable or permanent sealants
  • Seals to match most microplate materials and applications, including polypropylene, polystyrene and COC
Agilent offers a range of PlateLoc microplate heat seals made from aluminum-based or optically clear materials with peelable, pierceable or permanent sealants matching most microplate materials and applications.
Ideal for life science applications that require automated microplate heat sealing, Agilent heat seals keep the PlateLoc Thermal Microplate Sealer up and running reliably to securely heat seal your microplates.