Imprimer
Microplate Lids and Seals
Fournisseur: AGILENT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
200856-100 202497-100
AGLS200856-100EA 137 EUR
AGLS200856-100 AGLS202497-100
Microplate Lids and Seals
Couvercles Couvercles pour plaques
Universal lids and cap mats for square microplates.
  • Universal-sized polystyrene lids fit most all microplates
  • Black colour is effective for light-sensitive reagents
  • Robot-friendly design
  • Cap mats specifically designed for optimal fit for Agilent square-well microplates
  • Mat material has low permeability to alcohols
Agilent microplate accessories include microplate lids and cap mats. Agilent supplies universal lids for microplates, available in clear or black polystyrene resin. Custom cap mats are available for sealing Agilent 24, 48, and 96-square well microplates. These custom cap mats are molded in Santoprene, a chemical-resistant thermoplastic elastomer.