Imprimer
Disposable Tip Pipette Heads
Fournisseur: AGILENT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
G5055G G5056G G5057G
AGLSG5055GEA 41659 EUR
AGLSG5055G AGLSG5056G AGLSG5057G
Disposable Tip Pipette Heads
Pointes de pipettes
Agilent pipette heads are designed to be easily mounted or exchanged and to offer the advantage of being more forgiving than metal-tipped, fixed-tip heads during collisions, increasing uptime during the life of your instrument.
  • Designed to work in two liquid volume ranges: small transfer (ST) and large transfer (LT)
  • Typical %CVs for dispensing 2 µl are 2,1% using DMSO
  • Compatible with most common life science laboratory reagents and, with optimisation, CVs better than 5% can be achieved across many liquid types
Disposable-tip pipette heads can aspirate or dispense into one sample well or an array of sample wells (which include a corner and are contiguous). This is primarily used for single-tip cherry picking and occasionally for multiple-tip or array cherry picking.
Agilent disposable-tip pipette heads and tips cover a pipetting volume range from 300 nl to 250 µl with 5% CV. High precision is achieved by optimising each head for a specific portion of the pipetting volume range.