Imprimer
Reservoir Microplates
Fournisseur: AGILENT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
201254-100 201244-100 200686-100 204363-100 204484-100 201250-100 204093-100 201252-100 204504-100 204017-100 201246-100 201256-100 204095-100 201280-100 204365-100 201284-100 204284-100
AGLS201254-100EA 155 EUR
AGLS201254-100 AGLS201244-100 AGLS200686-100 AGLS204363-100 AGLS204484-100 AGLS201250-100 AGLS204093-100 AGLS201252-100 AGLS204504-100 AGLS204017-100 AGLS201246-100 AGLS201256-100 AGLS204095-100 AGLS201280-100 AGLS204365-100 AGLS201284-100 AGLS204284-100
Reservoir Microplates
Microplaques Microplaques HTS
Polypropylene reservoir microplates for automated liquid handlers and handheld pipettors.
  • Large range of cavity volumes, from 2,6 to 300 ml with specialised bottom geometries to optimise access and minimise dead volumes
  • Low binding surfaces for minimal losses of proteins, DNA/RNA, and other biologicals
  • Molded from chemical-resistant polymers, for maintained performance following prolonged exposure to aggressive solutions and solvents
  • Produced to ANSI/SLAS standards, for compatibility with popular labware and devices
  • Compatible with all major platforms and benchtop devices, making these reservoirs automation friendly
  • Low temperature tolerant, suitable for heating or cooling during use
  • Autoclavable, can be sterilised at 121 °C, 1,03 bar for 20 minutes
  • Fully customisable to fit your requirements
  • Manufactured in a DNAse/RNase-free ISO 9001 regulated facility
Agilent polypropylene reservoir microplates are available for a wide range of automated liquid handlers and handheld pipettors. These reservoir microplates are molded using high-quality, chemical-resistant polymers to ANSI/SLAS standard footprints, with optimised base geometries to minimise dead volumes. They are available with 1 to 24 cavities per reservoir plate, container heights between 19 and 44 mm, and with clear or black lids.
These reservoirs are recommended for both method scale-up and scale-down applications as well as for creating design of experiments (DoE) arrays.