Imprimer
LABMAX Premium Bottle-Top Dispensers
Fournisseur: witeg Labortechnik
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
5.370.904 5.370.906 5.370.905
WITG5.370.904EA 555 EUR
WITG5.370.904 WITG5.370.906 WITG5.370.905
LABMAX Premium Bottle-Top Dispensers
Distributeurs pour flacons
The universal dispenser LABMAX premium has a sophisticated system which allows air purging and draining without any loss of reagent. Due to the direct displacement system, LABMAX premium is ideal for crystalline solutions. We improved the adjust knob for a quick and precise volume setting. On top, we added the built-in recalibration mechanism.
  • Easy to clean, completely autoclavable at 121 °C
  • Highest accuracy and precision
  • All-purpose dispenser, universal chemical compatibility
  • Direct displacement piston system, ideal for crystalline solutions
  • Built-in recalibration mechanism including calibration tool
  • Robust and stable, made of high quality materials e.g. platinum springs
  • One handed operation and flexible by using different bottle adapters
0% Loss of reagent by pumping in a closed circuit, 100% drip-free due to our unique anti-drip system. 360° rotatable valve block for added safety with 2 scales - for measurements in milliliter and ounces. Telescopic suction tube included in scope of supply.
Informations de livraison: Supplied with telescopic suction tube, discharge tube, instruction manual, calibration tool, individual certificate, serial number, thread adapter.