Imprimer
Urine Cups with Integrated Transfer Device
Fournisseur: Greiner Bio-One
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
724321 724322
GREI724321EA 185 EUR
GREI724321 GREI724322
Urine Cups with Integrated Transfer Device
Systèmes de diagnostic clinique Prélèvement/préparation d'échantillons de diagnostic clinique Prélèvement/préparation d'échantillons d'urine
The Urine Cup with integrated transfer device allows you to collect and transport urine samples safely to the necessary destination. Thanks to the integrated transfer device, sample transfer from the cup to an evacuated tube is simple, fast and hygienic.
  • Recessed transfer device to reduce risk of needle stick injuries
  • Available bulk packed or in cases of small packs to suit different distribution needs within healthcare facilities
The optimised threading of the urine cup makes it easy to open and close. In addition, a stabilising flange makes the cup closure more secure and prevents over-threading.
The perforated label acts as a sterility integrity seal visually indicating if the product has been opened prior to use.