Imprimer

Anti-Syndecan-1 Rat Monoclonal Antibody [clone: 281-2] (Biotin)

Fournisseur: STEMCELL Technologies
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

60035BT 60035BT.1
STMC60035BTEA 704 EUR
STMC60035BT STMC60035BT.1
Anti-Syndecan-1 Rat Monoclonal Antibody [clone: 281-2] (Biotin)
Anticorps
Rat monoclonal IgG2a antibody against mouse CD138 (syndecan-1), biotin-conjugated.

  • B cells
  • Plasma

The 281-2 antibody reacts with an extracellular epitope on the core protein of mouse CD138 (Syndecan-1), a ~60 to 100 kDa type 1 transmembrane glycoprotein expressed on the surface of pre-B cells, immature B cells, and normal and malignant plasma cells (but not mature circulating B cells). CD138 is also expressed on non-hematopoietic cells such as embryonic mesenchymal cells, endothelial, epithelial and neural cells. CD138 is thought to act primarily as a receptor that modulates cell proliferation, cell migration and cell-matrix associations by linking the extracellular matrix to the cytoskeleton. Heparin sulfate and chondroitin sulfate moieties attached to CD138 associate with several proteins, including collagens, fibronectin, tenascin, thrombospondin and certain cytokines.

Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type: Anti-Syndecan-1 Rat Monoclonal Antibody [clone: 281-2] (Biotin)
Antigen: Syndecan-1
Clonality: Monoclonal
Clone: 281-2
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR