Imprimer

Anti-CD49b Rat Monoclonal Antibody [clone: DX5] (FITC)

Fournisseur: STEMCELL Technologies
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

60020FI 60020FI.1
STMC60020FIEA 0 EUR
STMC60020FI STMC60020FI.1
Anti-CD49b Rat Monoclonal Antibody [clone: DX5] (FITC)
Anticorps
Rat monoclonal IgM antibody against mouse CD49b (integrin α2), FITC-conjugated.

  • Nk cells

The DX5 antibody reacts with murine CD49b (integrin α2), an ~150 kDa type 1 transmembrane glycoprotein that associates non-covalently with CD29 (integrin β1) to form the heterodimeric CD49b/CD29 complex known as VLA-2, a receptor for extracellular matrix proteins such as collagen, E-cadherin, fibronectin, and laminin. CD49b is highly expressed by platelets and is found on a majority of NK cells, on NKT cells, and on a small subset of CD8+ T cells; the latter population increases substantially following viral infection. CD49b is also expressed by several tissues, including intestine, kidney, mammary gland, and lung. The DX5 antibody is particularly useful for identifying NK cells in mice lacking the NK1.1 antigen. Binding of the DX5 antibody has not been observed to affect the function of the VLA-2 integrin. DX5 binding is, however, blocked by the clone HMα2 antibody. This antibody clone has been verified for purity assessments of cells isolated with EasySep™ Mouse NK Cell Enrichment Kit (Catalog #19755).

Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: CD49b
Clonality: Monoclonal
Clone: DX5
Conjugation: FITC
Epitope:
Host: Rat
Isotype: IgM kappa
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR