Imprimer
HypoThermosol® FRS Preservation Medium
Marques : STEMCELL Technologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
07935 07936 07945 07934
STMC07935EA 380 EUR
STMC07935 STMC07936 STMC07945 STMC07934
HypoThermosol® FRS Preservation Medium
Milieux de cryoconservation cellulaire
Animal component-free, defined hypothermic (2 to 8 °C) preservation medium for a range of cell and tissue types.
  • Ready-to-use
  • Serum-free, protein-free
  • Animal component-free
  • cGMP manufactured with USP grade / highest-quality components
  • FDA master file
  • Sterility, endotoxin, and cell-based release testing
Improve and extend the preservation of cells, tissues, and organs with HypoThermosol® FRS. This ready-to-use, serum-free, protein-free, and animal component-free preservation medium is formulated to address the molecular-biological response of cells during the hypothermic (2 to 8°C) preservation process. HypoThermosol® FRS includes key ions at concentrations that balance the intracellular state at hypothermic temperatures. This medium has been subjected to sterility, endotoxin, and cell-based release testing and is manufactured under cGMPs with USP-grade components. Additional components include pH buffers, energy substrates, free radical scavengers, and osmotic/oncotic stabilisers.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').