Imprimer
Ammonium Chloride Solution
Marques : STEMCELL Technologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
07850 07800
STMC07850EA 259 EUR
STMC07850 STMC07800
Ammonium Chloride Solution
Tampons Tampons d'extraction
Ammonium chloride solution is recommended for the lysis of red blood cells (RBCs) in preparations of human and mouse peripheral blood, spleen or bone marrow cells. It is buffered and optimised for gentle lysis of erythrocytes, with minimal effect on leukocytes. It does not contain a fixative agent, therefore leukocytes are viable following RBC lysis.
Species: Human, mouse, non human primate, other, rat
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').