Imprimer
Test strips, water hardness, AQUADUR®
Marques : MACHEREY-NAGEL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
91220 91223 91221 91224 912902 91201 91239 91240
662-0239EA 47.8 EUR
662-0239 MANA91223 MANA91221 MANA91224 MANA912902 662-1867 MANA91239 MANA91240
Test strips, water hardness, AQUADUR®
Kits de tests environnementaux
AQUADUR® test strips are for the quick and easy determination of water hardness. The convenient dip and read procedure and the clear colour changes ensure reliable results within seconds.
  • No accessories needed, always ready to use
  • Quick and easy determination of the water hardness
  • Easy read-off due to brilliant colour scale
The water hardness depends mainly on the amount of Calcium and Magnesium salts within the water. The total sum of these salts determines the actual water hardness. The water hardness is expressed in ppm (mg/L) CaCO₃. AQUADUR® water hardness test strips are the ideal choice to quickly, easily and reliably determine water hardness.
AQUADUR® water hardness test strips come in a wide variety of different gradations and packaging options for individual needs. Single sealed test strips are for example available to inform customers about the necessity of water softeners.