Imprimer
SteriWare® Scrapers, Blue
Fournisseur: Sampling Systems
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
8785B-75 8785B-75S
SAMP8785B-75EA 354 EUR
SAMP8785B-75 SAMP8785B-75S
SteriWare® Scrapers, Blue
Raclettes
The SteriWare scraper is designed for use in production or laboratory applications.
  • Available in non-sterile or sterile formats
  • 75 mm blade width
  • Used for scraping, removing and splitting product
  • Available from stock
  • Fast delivery
These blue disposable scrapers are ideal for use in multiple applications, such as removing excess product or scraping machines clean after use. The bright blue colour allows it to be easily spotted if left in material.