Imprimer
Sampling Scoop and Lid
Fournisseur: Sampling Systems
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
8916A-100 8916A-100S 8916A-50S 8916A-50 8916A-25S 8916A-25
SAMP8916A-100EA 228 EUR
SAMP8916A-100 SAMP8916A-100S SAMP8916A-50S SAMP8916A-50 SAMP8916A-25S SAMP8916A-25
Sampling Scoop and Lid
Scoops
Sampling scoops with lid are designed for easy sample handling.
  • Scoop comes with clear, snap-on lid
  • Avoids the need for additional sample containers
  • Flat bottom for easy weighing
  • Sterile and non sterile versions available
  • Available from stock
  • Fast delivery
This range of scoops with lids are the perfect answer to reducing sampling errors. Simply snap on the clear lid and collect samples straight for analysis without the need to transfer to any additional containers. The flat bottom design makes it easy to weigh the sample.