Imprimer
Jumbo Sack Sampler
Fournisseur: Sampling Systems
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1309C-400 1309C-800 1309B-400 1309B-800 1309A-400 1309A-800
SAMP1309C-400EA 702 EUR
SAMP1309C-400 SAMP1309C-800 SAMP1309B-400 SAMP1309B-800 SAMP1309A-400 SAMP1309A-800
Jumbo Sack Sampler
Échantillonneurs Préleveurs de solides
The jumbo sack sampler is designed for sampling large granules from sacks.
  • Ideal for sampling granules from bags on pallets
  • Can sample larger granules up to 10mm in size
  • Simple to use
  • Ideal for most powders and granules
  • Available from stock
  • Fast delivery
The jumbo aack sampler is made from 316L Stainless steel, and is perfect for sampling powders and granules in sacks. There are various models to suit specific applications, including a model with a plain end which the user can attach to their own bag. A model with an end cap allows the user to retain the sample in the tube. Additionally, a model is available with an adaptor to enable the user to collect the sample in a bottle.